Merck
  • Modulation of the multixenobiotic resistance mechanism in Danio rerio hepatocyte culture (ZF-L) after exposure to glyphosate and Roundup®.
Modulation of the multixenobiotic resistance mechanism in Danio rerio hepatocyte culture (ZF-L) after exposure to glyphosate and Roundup®.
Chemosphere (2019-04-29)
Lopes Fernanda Moreira, Sandrini Juliana Zomer, Souza Marta Marques
ABSTRACT
The presence of the transmembrane proteins of the ATP-binding cassette (ABC) family, which perform the efflux of several substances, contributes to the survival of aquatic organisms in a contaminated environmental. Those proteins provide a phenotype named the multixenobiotic resistance mechanism (MXR) by performing the efflux of a wide range of endogenous and exogenous compounds (ABCB) and biotransformation products and anionic compounds (ABCC). The aim of the present study was to evaluate the cellular defense pathway of an established culture from zebrafish hepatocytes (ZF-L) after 24 and 48 h of exposure to glyphosate and Original Roundup®, an herbicide used globally. Through abcb4, abcc1, abcc2 and abcc4 gene expression, ABCB and ABCC2 protein expression and ABC pump activity in ZF-L cells exposed to glyphosate and Roundup®. The results showed an increase in ABCB gene and protein expression; however, although ABCC2 showed an increase in gene expression, its protein expression was lower than in the control group. Regarding ABC activity, only exposure to Roundup® at the lowest concentration showed an increase at 48 h, but in the presence of inhibitors, both glyphosate and Roundup® appeared to modulate ABC activity, reducing its inhibition and returning activity to levels without inhibitor.