Merck
  • Characterization of Double-Doped Polymer Optical Fibers as Luminescent Solar Concentrators.
Characterization of Double-Doped Polymer Optical Fibers as Luminescent Solar Concentrators.
Polymers (2019-07-18)
Itxaso Parola, M Asuncion Illarramendi, Florian Jakobs, Jana Kielhorn, Daniel Zaremba, Hans-Hermann Johannes, Joseba Zubia
ABSTRACT
This work reports on a diameter dependence analysis of the performance as luminescent solar concentrators of three self-fabricated polymer optical fibers (POFs) doped with a hybrid combination of dopants. The works carried out include the design and self-fabrication of the different diameter fibers; an experimental analysis of the output power, of the output irradiance and of the fluorescent fiber solar concentrator efficiency; a comparison of the experimental results with a theoretical model; a study of the performance of all the fibers under different simulated lighting conditions; and a calculation of the active fiber length of each of the samples, all of them as a function of the fiber core diameter. To the best of our knowledge, this paper reports the first analysis of the influence of the POF diameter for luminescent solar concentration applications. The results obtained offer a general perspective on the optimal design of solar energy concentrating systems based on doped POFs and pave the way for the implementation of cost-effective solar energy concentrating devices.