Merck
  • Diversity and convergence of mechanisms involved in pyrethroid resistance in the stored grain weevils, Sitophilus spp.
Diversity and convergence of mechanisms involved in pyrethroid resistance in the stored grain weevils, Sitophilus spp.
Scientific reports (2018-11-07)
Khalid Haddi, Wilson R Valbon, Luis O Viteri Jumbo, Luiz O de Oliveira, Raul N C Guedes, Eugenio E Oliveira
ABSTRACT
Target-site mutations and changes in insect metabolism or behavior are common mechanisms in insecticide-resistant insects. The co-occurrence of such mechanisms in a pest strain is a prominent threat to their management, particularly when alternative compounds are scarce. Pyrethroid resistance among stored grain weevils (i.e., Sitophilus spp.) is an example of a long-standing concern, for which reports of resistance generally focus on a single mechanism in a single species. Here, we investigated pyrethroid resistance in maize and rice weevils (i.e., Sitophilus zeamais and S. oryzae), exploring potential knockdown resistance (kdr) mutations in their sodium channels (primary site for pyrethroid actions) and potential changes in their detoxification and walking processes. Resistance in pyrethroid-resistant rice weevils was associated with the combination of a kdr mutation (L1014F) and increases in walking and detoxification activities, while another kdr mutation (T929I) combined with increases in walking activity were the primary pyrethroid resistance mechanisms in maize weevils. Our results suggest that the selection of pyrethroid-resistant individuals in these weevil species may result from multiple and differential mechanisms because the L1014F mutation was only detected in Latin American rice weevils (e.g., Brazil, Argentina and Uruguay), not in Australian and Turkish rice weevils or Brazilian maize weevils.