Merck
  • Selective inhibition of farnesyl-protein transferase blocks ras processing in vivo.
Selective inhibition of farnesyl-protein transferase blocks ras processing in vivo.
The Journal of biological chemistry (1993-04-15)
J B Gibbs, D L Pompliano, S D Mosser, E Rands, R B Lingham, S B Singh, E M Scolnick, N E Kohl, A Oliff
ABSTRACT
The ras oncogene product, Ras, is synthesized in vivo as a precursor protein that requires post-translational processing to become biologically active and to be capable of transforming mammalian cells. Farnesylation appears to be a critical modification of Ras, and thus inhibitors of the farnesyl-protein transferase (FPTase) that catalyzes this reaction may block ras-dependent tumorigenesis. Three structural classes of FPTase inhibitors were identified: (alpha-hydroxyfarnesyl)phosphonic acid, chaetomellic acids, and zaragozic acids. By comparison, these compounds were weaker inhibitors of geranylgeranyl-protein transferases. Each of these inhibitors was competitive with respect to farnesyl diphosphate in the FPTase reaction. All compounds were assayed for inhibition of Ras processing in Ha-ras-transformed NIH3T3 fibroblasts. Ras processing was inhibited by 1 microM (alpha-hydroxyfarnesyl)phosphonic acid. Neither chaetomellic acid nor zaragozic acid were active in this assay. These results are the first demonstration that a small organic chemical selected for inhibition of FPTase can inhibit Ras processing in vivo.