Merck
  • Discovery of a new series of Aurora inhibitors through truncation of GSK1070916.
Discovery of a new series of Aurora inhibitors through truncation of GSK1070916.
Bioorganic & medicinal chemistry letters (2010-03-26)
Jesus R Medina, Seth W Grant, Jeffrey M Axten, William H Miller, Carla A Donatelli, Mary Ann Hardwicke, Catherine A Oleykowski, Qiaoyin Liao, Ramona Plant, Hong Xiang
ABSTRACT
Novel Aurora inhibitors were identified truncating clinical candidate GSK1070916. Many of these truncated compounds retained potent activity against Aurora B with good antiproliferative activity. Mechanistic studies suggested that these compounds, depending on the substitution pattern, may or may not exert their antiproliferative effects via inhibition of Aurora B. The SAR results from this investigation will be presented with an emphasis on the impact structural changes have on the cellular phenotype.
Sigma-Aldrich
4-Pyrazoleboronic acid pinacol ester, 97%