Merck
  • Phosphorylation of histone H3.3 at serine 31 promotes p300 activity and enhancer acetylation.
Phosphorylation of histone H3.3 at serine 31 promotes p300 activity and enhancer acetylation.
Nature genetics (2019-06-04)
Sara Martire, Aishwarya A Gogate, Amanda Whitmill, Amanuel Tafessu, Jennifer Nguyen, Yu-Ching Teng, Melodi Tastemel, Laura A Banaszynski
ABSTRACT
The histone variant H3.3 is enriched at enhancers and active genes, as well as repeat regions such as telomeres and retroelements, in mouse embryonic stem cells (mESCs)1-3. Although recent studies demonstrate a role for H3.3 and its chaperones in establishing heterochromatin at repeat regions4-8, the function of H3.3 in transcription regulation has been less clear9-16. Here, we find that H3.3-specific phosphorylation17-19 stimulates activity of the acetyltransferase p300 in trans, suggesting that H3.3 acts as a nucleosomal cofactor for p300. Depletion of H3.3 from mESCs reduces acetylation on histone H3 at lysine 27 (H3K27ac) at enhancers. Compared with wild-type cells, those lacking H3.3 demonstrate reduced capacity to acetylate enhancers that are activated upon differentiation, along with reduced ability to reprogram cell fate. Our study demonstrates that a single amino acid in a histone variant can integrate signaling information and impact genome regulation globally, which may help to better understand how mutations in these proteins contribute to human cancers20,21.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Histone H3.1/H3.2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser28) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Daxx Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Histone H3.3 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-acetyl Histone H3 Antibody (Lys 64), from rabbit, purified by affinity chromatography