Merck
  • Free-standing supercapacitors from Kraft lignin nanofibers with remarkable volumetric energy density.
Free-standing supercapacitors from Kraft lignin nanofibers with remarkable volumetric energy density.
Chemical science (2019-04-19)
Philipp Schlee, Servann Herou, Rhodri Jervis, Paul R Shearing, Dan J L Brett, Darren Baker, Omid Hosseinaei, Per Tomani, M Mangir Murshed, Yaomin Li, María José Mostazo-López, Diego Cazorla-Amorós, Ana Belen Jorge Sobrido, Maria-Magdalena Titirici
ABSTRACT
We have discovered a very simple method to address the challenge associated with the low volumetric energy density of free-standing carbon nanofiber electrodes for supercapacitors by electrospinning Kraft lignin in the presence of an oxidizing salt (NaNO3) and subsequent carbonization in a reducing atmosphere. The presence of the oxidative salt decreases the diameter of the resulting carbon nanofibers doubling their packing density from 0.51 to 1.03 mg cm-2 and hence doubling the volumetric energy density. At the same time, the oxidative NaNO3 salt eletrospun and carbonized together with lignin dissolved in NaOH acts as a template to increase the microporosity, thus contributing to a good gravimetric energy density. By simply adjusting the process parameters (amount of oxidizing/reducing agent), the gravimetric and volumetric energy density of the resulting lignin free-standing carbon nanofiber electrodes can be carefully tailored to fit specific power to energy demands. The areal capacitance increased from 147 mF cm-2 in the absence of NaNO3 to 350 mF cm-2 with NaNO3 translating into a volumetric energy density increase from 949 μW h cm-3 without NaNO3 to 2245 μW h cm-3 with NaNO3. Meanwhile, the gravimetric capacitance also increased from 151 F g-1 without to 192 F g-1 with NaNO3.