- MED28 and forkhead box M1 (FOXM1) mediate matrix metalloproteinase 2 (MMP2)-dependent cellular migration in human nonsmall cell lung cancer (NSCLC) cells.
MED28 and forkhead box M1 (FOXM1) mediate matrix metalloproteinase 2 (MMP2)-dependent cellular migration in human nonsmall cell lung cancer (NSCLC) cells.
Non-small-cell lung cancer (NSCLC) accounts for the majority of the lung cancer cases that have become a leading cause of cancer deaths worldwide. Overexpression of transcription factor forkhead box M1 (FOXM1) is involved in the inauspicious development of several types of cancer, including lung tumor aggressiveness. Our laboratory has previously found that MED28, a Mediator subunit for transcriptional activation, modulates cell growth, epithelial-mesenchymal transition, migration, and invasion in human breast cancer cells. The objective of the current study is to investigate the potential role of MED28 and FOXM1 in NSCLC. In addition to A549 and PC9 cells, we also used a doxycycline-inducible system to generate FOXM1-overexpressed A549-DN cells, and we explored the connection of MED28 with FOXM1 and their effect on migration. Herein, we report that the increased expression levels of both MED28 and FOXM1 elevated the expression of matrix metalloproteinase 2 (MMP2), a metastasis marker, which enhanced cell migration and matrigel invasion of NSCLC cells. Furthermore, MED28 interacted with FOXM1, and both exhibited a mutual effect on the expression and subcellular localization. Moreover, MED28 small interfering RNA-mediated MMP2 gene suppression could be attenuated by inducible expression of a constitutively active form of FOXM1, which consequently restored the migration and invasion ability of NSCLC cells. Our data indicate that MED28 interacts with FOXM1, and each affects the expression and localization of the other, and, more importantly, both regulate MMP2-dependent migration and invasion in human lung cancer cells.