Merck
  • Synthesis of new trisulfonated calix[4]arenes functionalized at the upper rim, and their complexation with the trimethyllysine epigenetic mark.
Synthesis of new trisulfonated calix[4]arenes functionalized at the upper rim, and their complexation with the trimethyllysine epigenetic mark.
Organic letters (2012-03-09)
Kevin D Daze, Manuel C F Ma, Florent Pineux, Fraser Hof
ABSTRACT
A synthetic route to produce a new family of trisulfonated calix[4]arenes bearing a single group, selectively introduced, that lines the binding pocket is reported. Ten examples, including new sulfonamide and biphenyl-substituted hosts, each with additional binding elements, demonstrate the tuning of guest affinities and selectivities. NMR titrations in phosphate-buffered water show that one of the new hosts binds to the modified amino acid trimethyllysine with the highest affinity and selectivity observed to date.
Sigma-Aldrich
4-Carboxyphenylboronic acid
Sigma-Aldrich
Nε,Nε,Nε-Trimethyllysine hydrochloride, ≥97% (TLC)
Sigma-Aldrich
4-Cyanophenylboronic acid, ≥95%