Merck
  • PTEN-induced partial epithelial-mesenchymal transition drives diabetic kidney disease.
PTEN-induced partial epithelial-mesenchymal transition drives diabetic kidney disease.
The Journal of clinical investigation (2019-02-12)
Yajuan Li, Qingsong Hu, Chunlai Li, Ke Liang, Yu Xiang, Heidi Hsiao, Tina K Nguyen, Peter K Park, Sergey D Egranov, Chandrashekar R Ambati, Nagireddy Putluri, David H Hawke, Leng Han, Mien-Chie Hung, Farhad R Danesh, Liuqing Yang, Chunru Lin
ABSTRACT
Epithelial-mesenchymal transition (EMT) contributes significantly to interstitial matrix deposition in diabetic kidney disease (DKD). However, detection of EMT in kidney tissue is impracticable, and anti-EMT therapies have long been hindered. We reported that phosphatase and tensin homolog (PTEN) promoted transforming growth factor beta 1 (TGF-β), sonic hedgehog (SHH), connective tissue growth factor (CTGF), interleukin 6 (IL-6), and hyperglycemia-induced EMT when PTEN was modified by a MEX3C-catalyzed K27-linked polyubiquitination at lysine 80 (referred to as PTENK27-polyUb). Genetic inhibition of PTENK27-polyUb alleviated Col4a3 knockout-, folic acid-, and streptozotocin-induced (STZ-induced) kidney injury. Serum and urine PTENK27-polyUb concentrations were negatively correlated with glomerular filtration rate (GFR) for diabetic patients. Mechanistically, PTENK27-polyUb facilitated dephosphorylation and protein stabilization of TWIST, SNAI1, and YAP in renal epithelial cells, leading to enhanced EMT. We identified that a small molecule, triptolide, inhibited MEX3C-catalyzed PTENK27-polyUb and EMT of renal epithelial cells. Treatment with triptolide reduced TWIST, SNAI1, and YAP concurrently and improved kidney health in Col4a3 knockout-, folic acid-injured disease models and STZ-induced, BTBR ob/ob diabetic nephropathy models. Hence, we demonstrated the important role of PTENK27-polyUb in DKD and a promising therapeutic strategy that inhibited the progression of DKD.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M1 antibody, clone M1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-UBE2S antibody produced in mouse, clone 2G7, purified immunoglobulin, buffered aqueous solution