Merck
  • A new target for Alzheimer's disease: A small molecule SERCA activator is neuroprotective in vitro and improves memory and cognition in APP/PS1 mice.
A new target for Alzheimer's disease: A small molecule SERCA activator is neuroprotective in vitro and improves memory and cognition in APP/PS1 mice.
Bioorganic & medicinal chemistry letters (2018-04-01)
Katherine Krajnak, Russell Dahl
ABSTRACT
Amongst the cellular cacophony of altered signals in Alzheimer's disease (AD), disrupted Ca2+ homeostasis and consequential endoplasmic reticulum (ER) stress signals have been recognized as key determinants of neuron fate. This altered Ca2+ state is accompanied by a failing sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) pump, which has been recognized as a causal feature of the underlying disease state. Repair of the Ca2+ dyshomeostasis represents a putative drug target via alleviation of ER stress and rescue of injured neurons, effectively modifying the AD state. Herein, we report a small molecule SERCA activator that rescues brain cells and raises ER Ca2+ in vitro, and shows efficacy in the APP/PS1 double transgenic mouse model of Alzheimer's disease. These results support SERCA activation as a therapeutic target for AD.
Sigma-Aldrich
CDN1163, ≥98% (HPLC)