Merck
  • The influence of sphingomyelin on the structure and function of reconstituted high density lipoproteins.
The influence of sphingomyelin on the structure and function of reconstituted high density lipoproteins.
The Journal of biological chemistry (1996-02-23)
K A Rye, N J Hime, P J Barter
ABSTRACT
The effect of sphingomyelin (SPM) on the structure and function of discoidal and spherical reconstituted high density lipoproteins (rHDL) has been studied. Three preparations of discoidal rHDL with 1-palmitoyl-2-oleoyl phosphatidylcholine (POPC)/SPM/unesterified cholesterol (UC)/apolipoprotein (apo)A-I molar ratios of 99.6/0. 0/10.2/1.0, 86.0/13.6/10.8/1.0, and 72.5/26.3/11.4/1.0 were prepared by cholate dialysis. SPM did not affect discoidal rHDL size or surface charge. Esterification of cholesterol by lecithin:cholesterol acyltransferase (LCAT) was inhibited in the SPM-containing discoidal rHDL. When the discoidal rHDL of POPC/SPM/UC/apoA-I molar ratio 99.6/0.0/10.2/1.0 were incubated with low density lipoproteins (LDL) and LCAT, SPM transferred spontaneously from the LDL to the rHDL (t1/2 = 0.8 h) and spherical particles with a POPC/SPM/UC/CE/apoA-I molar ratio of 24.6/4.9/3. 6/24.9/1.0 were formed. Depleting the spherical rHDL of SPM head groups by incubation with sphingomyelinase increased the negative charge on the surface, but did not change their size. Cholesteryl ester transfer protein (CETP)-mediated transfers of cholesteryl esters and triglyceride between spherical rHDL and Intralipid were not affected by SPM head group depletion. The effect of SPM on rHDL structure was assessed spectroscopically. SPM increased POPC acyl chain and head group packing in the discoidal rHDL. When the spherical rHDL were depleted of SPM head groups, POPC acyl chain packing order decreased, but head group packing order was not affected. SPM inhibited the lipid-water interfacial hydration of discoidal rHDL. This parameter was not affected when the spherical rHDL were depleted of SPM head groups. The SPM molecule and the SPM head group, respectively, inhibited the unfolding of apoA-I in discoidal and spherical rHDL. It is concluded that (i) SPM influences the structure of discoidal and spherical rHDL, (ii) SPM inhibits the LCAT reaction in discoidal rHDL, and (iii) the SPM head group does not affect CETP-mediated lipid transfers into or out of spherical rHDL.