Merck
  • Nonradioactive quantification of autophagic protein degradation with L-azidohomoalanine labeling.
Nonradioactive quantification of autophagic protein degradation with L-azidohomoalanine labeling.
Nature protocols (2017-01-13)
Jigang Wang, Jianbin Zhang, Yew Mun Lee, Shukie Ng, Yin Shi, Zi-Chun Hua, Qingsong Lin, Han-Ming Shen
ABSTRACT
At present, several assays that use radioisotope labeling to quantify the degradation of long-lived proteins have been developed to measure autophagic flux. Here, we describe a nonradioactive pulse-chase protocol using L-azidohomoalanine (AHA) labeling to quantify long-lived protein degradation during autophagy. AHA is used as a surrogate for L-methionine, and, when added to cultured cells grown in methionine-free medium, AHA is incorporated into proteins during de novo protein synthesis. After a chase period to remove short-lived proteins, autophagy is induced by starvation or other stimuli. Cells then undergo a 'click' reaction between the azide group of AHA and a fluorescently tagged alkyne probe. The AHA-containing proteins can then be detected by flow cytometry. This protocol is nonradioactive, sensitive and quantitative, and it is easy to perform. It is also applicable to various cell culture systems. The whole protocol is estimated to take 4-5 d to complete.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
PP242 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture