Merck
  • The effects of aerobic, resistance, and combined exercise on metabolic control, inflammatory markers, adipocytokines, and muscle insulin signaling in patients with type 2 diabetes mellitus.
The effects of aerobic, resistance, and combined exercise on metabolic control, inflammatory markers, adipocytokines, and muscle insulin signaling in patients with type 2 diabetes mellitus.
Metabolism: clinical and experimental (2011-03-08)
Maria Luiza Mendonça Pereira Jorge, Vanessa Neves de Oliveira, Nathalia Maria Resende, Lara Ferreira Paraiso, Antonio Calixto, Angelica Lemos Debs Diniz, Elmiro Santos Resende, Eduardo Rochete Ropelle, Jose Barreto Carvalheira, Foued Salmen Espindola, Paulo Tannus Jorge, Bruno Geloneze
ABSTRACT
The purpose of this study was to compare the effects of 3 different modalities of exercise on metabolic control, insulin resistance, inflammatory markers, adipocytokines, and tissue expression of insulin receptor substrate (IRS)-1 after 12 weeks of training among patients with type 2 diabetes mellitus. Forty-eight patients with type 2 diabetes mellitus were randomly assigned to 4 groups of training (3 times a week, 60 minutes per session): aerobic group (n = 12), resistance group (n = 12), combined (aerobic and resistance) group (n = 12), and control group (n = 12). Fasting and postprandial blood glucose, glycated hemoglobin, lipid profile, insulin resistance index (homeostasis model assessment of insulin resistance), adipocytokines (adiponectin, visfatin, and resistin), tumor necrosis factor, interleukin, and high-sensitivity C-reactive protein (hs-CRP) were measured at baseline and at the end of the study. Patients also underwent a muscle microbiopsy before and after training to quantify IRS-1 expression. All 4 groups displayed decreases in blood pressure, fasting plasma glucose, postprandial plasma glucose, lipid profile, and hs-CRP (P < .05); and there was no difference across the groups. After training, the IRS-1 expression increased by 65% in the resistance group (P < .05) and by 90% in the combined group (P < .01). Exercise training favorably affects glycemic parameters, lipid profile, blood pressure, and hs-CRP. In addition, resistance and combined training can increase IRS-1 expression.
Product Description
Millipore
Human Insulin ELISA, This Human Insulin ELISA is used to measure & quantify insulin levels in Metabolism & Endocrine research.
Pricing and availability is not currently available.