Merck
  • Ara-ATP impairs 3'-end processing of pre-mRNAs by inhibiting both cleavage and polyadenylation.
Ara-ATP impairs 3'-end processing of pre-mRNAs by inhibiting both cleavage and polyadenylation.
Nucleic acids research (1991-11-11)
K Ghoshal, S T Jacob
ABSTRACT
Previous studies have demonstrated that Ara-ATP can inhibit poly(A) polymerase activity by competing with ATP. To elucidate the mechanism of action of this compound, its effect on the cleavage and polyadenylation of two specific substrates, SV40L and adenovirus L3 pre-mRNAs, was studied in HeLa nuclear extracts. Unlike cordycepin 5' triphosphate, Ara-ATP inhibited both cleavage and poly(A) addition. Addition of poly(A) polymerase fraction devoid of any other factors required for the processing reactions overcame the inhibitory effect on cleavage as well as polyadenylation of pre-mRNAs. These data suggest that Ara-ATP inhibits both cleavage and polyadenylation reactions by interacting with the ATP-binding site on poly(A) polymerase, the activity of which is essential for the cleavage reaction. Ara-ATP also blocked formation of the post-cleavage and polyadenylation-specific complexes, which further confirmed the inhibitory effect of the ATP analog on the two tightly coupled 3'-end processing reactions.
Sigma-Aldrich
Cordycepin 5′-triphosphate sodium salt, ≥95%