Merck
  • Regulation of integrin endocytic recycling and chemotactic cell migration by syntaxin 6 and VAMP3 interaction.
Regulation of integrin endocytic recycling and chemotactic cell migration by syntaxin 6 and VAMP3 interaction.
Journal of cell science (2012-05-11)
Krista A Riggs, Nazarul Hasan, David Humphrey, Christy Raleigh, Chris Nevitt, Deborah Corbin, Chuan Hu
ABSTRACT
Integrins are the primary receptors of cells adhering to the extracellular matrix, and play key roles in various cellular processes including migration, proliferation and survival. The expression and distribution of integrins at the cell surface is controlled by endocytosis and recycling. The present study examines the function of syntaxin 6 (STX6), a t-SNARE located in the trans-Golgi network, in integrin trafficking. STX6 is overexpressed in many types of human cancer. We show that depletion of STX6 inhibits chemotactic cell migration and the delivery of the laminin receptor α3β1 integrin to the cell surface, whereas STX6 overexpression stimulates chemotactic cell migration, integrin delivery, and integrin-initiated activation of focal adhesion kinase. These data indicate that STX6 plays a rate-limiting role in cell migration and integrin trafficking. In STX6-depleted cells, α3β1 integrin is accumulated in recycling endosomes that contain the v-SNARE VAMP3. Importantly, we show that STX6 and VAMP3 form a v-/t-SNARE complex, VAMP3 is required in α3β1 integrin delivery to the cell surface, and endocytosed α3β1 integrin traffics to both VAMP3 and STX6 compartments. Collectively, our data suggest a new integrin trafficking pathway in which endocytosed integrins are transported from VAMP3-containing recycling endosomes to STX6-containing trans-Golgi network before being recycled to the plasma membrane.
Sigma-Aldrich
Anti-Integrin α5β1 Antibody, clone HA5, clone HA5, Chemicon®, from mouse
Sigma-Aldrich
Anti-FAK Antibody, clone 4.47, clone 4.47, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-Paxillin (Tyr31) Antibody, clone M102, clone M102, from mouse
Sigma-Aldrich
Anti-Integrin α3 Antibody, serum, Chemicon®