Merck
  • Experimental assessment of effects of antiproliferative drugs of drug-eluting stents on endothelial cells.
Experimental assessment of effects of antiproliferative drugs of drug-eluting stents on endothelial cells.
Cardiovascular revascularization medicine : including molecular interventions (2015-08-09)
Keiichiro Miura, Haruaki Nakaya, Yoshio Kobayashi
ABSTRACT
Late and very late stent thrombosis after drug-eluting stent implantation is a major concern. The present study evaluated difference in the effects of sirolimus, paclitaxel and zotarolimus on endothelial cells. Mouse endothelial cells were seeded in a 6-well plate. Cells were cultured with an antiproliferative drug at the expected concentrations for each well for 24 hours before making 3 scratch lines with a pipette tip. After a 4.5 hour incubation period, 3 reference scratch lines, vertically across the original scratch lines, were made in the same way. The experiment was repeated at least 6 times (6 plates). Measurements were performed at 9 crossings of each well. Wound healing ratio was calculated as 1-(distance of the first scratch/distance of the second scratch). % cell migration was calculated as (wound healing ratio at an expected drug concentration/wound healing ratio with no drug) × 100. Average % cell migration at 54 crossings of 6 plates was calculated. Paclitaxel inhibited cell migration in a concentration-dependent manner. On the other hand, concentration-dependent inhibition was not observed for sirolimus or zotarolimus. Sirolimus showed a stronger inhibitory effect on migration of endothelial cells compared to zotarolimus. The difference in the effect of antiproliferative drugs of drug-eluting stents on endothelial cells may be associated with relatively faster re-endothelialization of zotarolimus-eluting stent compared to the 1st generation DES.
Sigma-Aldrich
Zotarolimus, ≥95% (HPLC)