Merck
  • Comparison of angioscopic findings among second-generation drug-eluting stents.
Comparison of angioscopic findings among second-generation drug-eluting stents.
Journal of cardiology (2016-12-31)
Yuji Nishimoto, Yasunori Ueda, Ryuta Sugihara, Ayaka Murakami, Keisuke Ueno, Yasuharu Takeda, Akio Hirata, Kazunori Kashiwase, Yoshiharu Higuchi, Yoshio Yasumura
ABSTRACT
First-generation drug-eluting stents (DES) have reduced short-term stent failure as compared to bare-metal stents due to the inhibition of neointima hyperplasia, but instead increased the risk of very-late stent failure. Although better outcomes have been reported for second-generation DES than for first-generation DES, the difference in the angioscopic findings at 1-year follow-up has not been adequately elucidated among second-generation DES. Consecutive 161 patients who received angioscopic examination at 1 year after implantation of second-generation DES, i.e. Nobori biolimus-eluting stents (Terumo, Tokyo, Japan) (N-BES, n=25), Xience everolimus-eluting stents (Abbot Vascular, Santa Clara, CA, USA; X-EES, n=95), or Resolute zotarolimus-eluting stents (Resolute Integrity; Medtronic, Minneapolis, MN, USA; R-ZES, n=41), in de novo native coronary lesions were analyzed. Maximum neointima coverage grade (N-BES, 0.9±0.3; X-EES, 1.2±0.4; R-ZES, 1.5±0.5; p<0.001) was the highest in R-ZES and lowest in N-BES. Heterogeneity score was higher in R-ZES than in N-BES (N-BES, 0.8±0.4; X-EES, 0.9±0.4; R-ZES, 1.1±0.5; p=0.007). Maximum yellow color grade and prevalence of thrombus were not different. Multivariate analysis demonstrated that only stent type was associated with maximum neointima coverage grade; stent type and total stent length were associated with heterogeneity score; and stenting for acute coronary syndrome (ACS) and total stent length were associated with maximum yellow color grade. Neointima coverage and heterogeneity were mainly determined by stent type even among second-generation DES, while yellow color was determined mainly by whether target lesion was of ACS.
Sigma-Aldrich
Zotarolimus, ≥95% (HPLC)