Merck
  • Custom-built electrostatics and supplementary bonding in the design of reinforced Collagen-g-P(methyl methacrylate-co-ethyl acrylate)/ nylon 66 core-shell fibers.
Custom-built electrostatics and supplementary bonding in the design of reinforced Collagen-g-P(methyl methacrylate-co-ethyl acrylate)/ nylon 66 core-shell fibers.
Journal of the mechanical behavior of biomedical materials (2018-07-22)
Zahra Bazrafshan, George K Stylios
ABSTRACT
In this study, Acid Soluble Collagen-g-P(methyl methacrylate-co-ethyl acrylate) (CME) was synthesized to take advantage of the flexibility of the resulted branched polymer chains and the high density of their chain entanglement. The coaxial electrospinning technique was applied to study the effect of electrically and structurally varied materials on fiber formation and fiber morphology when CME and Nylon 66 were electrospun as core and shell respectively. By tailoring the electrostatic field, different fiber content was achieved. The effect of chain orientation and intermolecular forces between the polymeric chains was investigated in the formed fibers by measuring thermal and mechanical properties, hydration degree and degradability. This approach to in situ fiber formation is not restricted to biomedical but has potential end-uses in a variety of multi-functional applications.
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%