Merck
  • Zinc induces CDK5 activation and neuronal death through CDK5-Tyr15 phosphorylation in ischemic stroke.
Zinc induces CDK5 activation and neuronal death through CDK5-Tyr15 phosphorylation in ischemic stroke.
Cell death & disease (2018-08-31)
Qing-Zhang Tuo, Zhen-Yu Liuyang, Peng Lei, Xiong Yan, Yang-Ping Shentu, Jia-Wei Liang, Huan Zhou, Lei Pei, Yan Xiong, Tong-Yao Hou, Xin-Wen Zhou, Qun Wang, Jian-Zhi Wang, Xiao-Chuan Wang, Rong Liu
ABSTRACT
CDK5 activation promotes ischemic neuronal death in stroke, with the recognized activation mechanism being calpain-dependent p35 cleavage to p25. Here we reported that CDK5-Tyr15 phosphorylation by zinc induced CDK5 activation in brain ischemic injury. CDK5 activation and CDK5-Tyr15 phosphorylation were observed in the hippocampus of the rats that had been subjected to middle cerebral artery occlusion, both of which were reversed by pretreatment with zinc chelator; while p35 cleavage and calpain activation in ischemia were not reversed. Zinc incubation resulted in CDK5-Tyr15 phosphorylation and CDK5 activation, without increasing p35 cleavage in cultured cells. Site mutation experiment confirmed that zinc-induced CDK5 activation was dependent on Tyr15 phosphorylation. Further exploration showed that Src kinase contributed to zinc-induced Tyr15 phosphorylation and CDK5 activation. Src kinase inhibition or expression of an unphosphorylable mutant Y15F-CDK5 abolished Tyr15 phosphorylation, prevented CDK5 activation and protected hippocampal neurons from ischemic insult in rats. We conclude that zinc-induced CDK5-Tyr15 phosphorylation underlies CDK5 activation and promotes ischemic neuronal death in stroke.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Histone H1 Antibody, clone AE-4, clone AE-4, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H1 Antibody, clone 12D11, culture supernatant, clone 12D11, from mouse
Sigma-Aldrich
10mM ATP
Sigma-Aldrich
Histone H1 Protein, 20 mg, Purified bovine Histone H1 protein for use as a substrate in histone modification assays (HAT, HDAC, DNMT) and chromatin assembly studies.