Merck
  • Human cDNA encoding the muscle isoform of the phosphorylase kinase gamma subunit (PHKG1).
Human cDNA encoding the muscle isoform of the phosphorylase kinase gamma subunit (PHKG1).
Human genetics (1995-11-01)
M Wehner, M W Kilimann
ABSTRACT
Muscle glycogenosis caused by phosphorylase kinase (Phk) deficiency may lead to exercise intolerance, weakness and musculatur atrophy. The gene encoding the muscle isoform of the Phk gamma subunit (gamma M) is one of the candidate genes in which mutations responsible for this condition should be sought. Here, we report the cDNA sequence and the predicted primary structure of the human gamma M subunit.
Sigma-Aldrich
PHKG1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.