MilliporeSigma
  • TREM-1 multimerization is essential for its activation on monocytes and neutrophils.
TREM-1 multimerization is essential for its activation on monocytes and neutrophils.
Cellular & molecular immunology (2018-03-24)
Kevin Carrasco, Amir Boufenzer, Lucie Jolly, Helene Le Cordier, Guanbo Wang, Albert Jr Heck, Adelheid Cerwenka, Emilie Vinolo, Alexis Nazabal, Alexandre Kriznik, Pierre Launay, Sebastien Gibot, Marc Derive
ABSTRACT
The triggering receptor expressed on myeloid cells-1 (TREM-1) is a receptor expressed on innate immune cells. By promoting the amplification of inflammatory signals that are initially triggered by Toll-like receptors (TLRs), TREM-1 has been characterized as a major player in the pathophysiology of acute and chronic inflammatory diseases, such as septic shock, myocardial infarction, atherosclerosis, and inflammatory bowel diseases. However, the molecular events leading to the activation of TREM-1 in innate immune cells remain unknown. Here, we show that TREM-1 is activated by multimerization and that the levels of intracellular Ca2+ release, reactive oxygen species, and cytokine production correlate with the degree of TREM-1 aggregation. TREM-1 activation on primary human monocytes by LPS required a two-step process consisting of upregulation followed by clustering of TREM-1 at the cell surface, in contrast to primary human neutrophils, where LPS induced a rapid cell membrane reorganization of TREM-1, which confirmed that TREM-1 is regulated differently in primary human neutrophils and monocytes. In addition, we show that the ectodomain of TREM-1 is able to homooligomerize in a concentration-dependent manner, which suggests that the clustering of TREM-1 on the membrane promotes its oligomerization. We further show that the adapter protein DAP12 stabilizes TREM-1 surface expression and multimerization. TREM-1 multimerization at the cell surface is also mediated by its endogenous ligand, a conclusion supported by the ability of the TREM-1 inhibitor LR12 to limit TREM-1 multimerization. These results provide evidence for ligand-induced, receptor-mediated dimerization of TREM-1. Collectively, our findings uncover the mechanisms necessary for TREM-1 activation in monocytes and neutrophils.
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit