Merck
  • Functional Annotation of ESR1 Gene Fusions in Estrogen Receptor-Positive Breast Cancer.
Functional Annotation of ESR1 Gene Fusions in Estrogen Receptor-Positive Breast Cancer.
Cell reports (2018-08-09)
Jonathan T Lei, Jieya Shao, Jin Zhang, Michael Iglesia, Doug W Chan, Jin Cao, Meenakshi Anurag, Purba Singh, Xiaping He, Yoshimasa Kosaka, Ryoichi Matsunuma, Robert Crowder, Jeremy Hoog, Chanpheng Phommaly, Rodrigo Goncalves, Susana Ramalho, Raquel Mary Rodrigues Peres, Nindo Punturi, Cheryl Schmidt, Alex Bartram, Eric Jou, Vaishnavi Devarakonda, Kimberly R Holloway, W Victoria Lai, Oliver Hampton, Anna Rogers, Ethan Tobias, Poojan A Parikh, Sherri R Davies, Shunqiang Li, Cynthia X Ma, Vera J Suman, Kelly K Hunt, Mark A Watson, Katherine A Hoadley, E Aubrey Thompson, Xi Chen, Shyam M Kavuri, Chad J Creighton, Christopher A Maher, Charles M Perou, Svasti Haricharan, Matthew J Ellis
ABSTRACT
RNA sequencing (RNA-seq) detects estrogen receptor alpha gene (ESR1) fusion transcripts in estrogen receptor-positive (ER+) breast cancer, but their role in disease pathogenesis remains unclear. We examined multiple ESR1 fusions and found that two, both identified in advanced endocrine treatment-resistant disease, encoded stable and functional fusion proteins. In both examples, ESR1-e6>YAP1 and ESR1-e6>PCDH11X, ESR1 exons 1-6 were fused in frame to C-terminal sequences from the partner gene. Functional properties include estrogen-independent growth, constitutive expression of ER target genes, and anti-estrogen resistance. Both fusions activate a metastasis-associated transcriptional program, induce cellular motility, and promote the development of lung metastasis. ESR1-e6>YAP1- and ESR1-e6>PCDH11X-induced growth remained sensitive to a CDK4/6 inhibitor, and a patient-derived xenograft (PDX) naturally expressing the ESR1-e6>YAP1 fusion was also responsive. Transcriptionally active ESR1 fusions therefore trigger both endocrine therapy resistance and metastatic progression, explaining the association with fatal disease progression, although CDK4/6 inhibitor treatment is predicted to be effective.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Anti-Estrogen Receptor α Antibody, clone 60C, rabbit monoclonal, culture supernatant, clone 60C, from rabbit
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture