Merck
  • Intragenic transcriptional interference regulates the human immune ligand MICA.
Intragenic transcriptional interference regulates the human immune ligand MICA.
The EMBO journal (2018-04-13)
Da Lin, Thomas K Hiron, Christopher A O'Callaghan
ABSTRACT
Many human genes have tandem promoters driving overlapping transcription, but the value of this distributed promoter configuration is generally unclear. Here we show that MICA, a gene encoding a ligand for the activating immune receptor NKG2D, contains a conserved upstream promoter that expresses a noncoding transcript. Transcription from the upstream promoter represses the downstream standard promoter activity in cis through transcriptional interference. The effect of transcriptional interference depends on the strength of transcription from the upstream promoter and can be described quantitatively by a simple reciprocal repressor function. Transcriptional interference coincides with recruitment at the standard downstream promoter of the FACT histone chaperone complex, which is involved in nucleosomal remodelling during transcription. The mechanism is invoked in the regulation of MICA expression by the physiological inputs interferon-γ and interleukin-4 that act on the upstream promoter. Genome-wide analysis indicates that transcriptional interference between tandem intragenic promoters may constitute a general mechanism with widespread importance in human transcriptional regulation.
Sigma-Aldrich
Anti-phospho RNA Pol II (Ser5), clone 1H4B6 Antibody, clone 1H4B6, from rat
Sigma-Aldrich
4-Thiouridine, ≥98%