Merck
  • Recombinant vitronectin is a functionally defined substrate that supports human embryonic stem cell self-renewal via alphavbeta5 integrin.
Recombinant vitronectin is a functionally defined substrate that supports human embryonic stem cell self-renewal via alphavbeta5 integrin.
Stem cells (Dayton, Ohio) (2008-07-05)
Stefan R Braam, Laura Zeinstra, Sandy Litjens, Dorien Ward-van Oostwaard, Stieneke van den Brink, Linda van Laake, Franck Lebrin, Peter Kats, Ron Hochstenbach, Robert Passier, Arnoud Sonnenberg, Christine L Mummery
ABSTRACT
Defined growth conditions are essential for many applications of human embryonic stem cells (hESC). Most defined media are presently used in combination with Matrigel, a partially defined extracellular matrix (ECM) extract from mouse sarcoma. Here, we defined ECM requirements of hESC by analyzing integrin expression and ECM production and determined integrin function using blocking antibodies. hESC expressed all major ECM proteins and corresponding integrins. We then systematically replaced Matrigel with defined medium supplements and ECM proteins. Cells attached efficiently to natural human vitronectin, fibronectin, and Matrigel but poorly to laminin + entactin and collagen IV. Integrin-blocking antibodies demonstrated that alphaVbeta5 integrins mediated adhesion to vitronectin, alpha5beta1 mediated adhesion to fibronectin, and alpha6beta1 mediated adhesion to laminin + entactin. Fibronectin in feeder cell-conditioned medium partially supported growth on all natural matrices, but in defined, nonconditioned medium only Matrigel or (natural and recombinant) vitronectin was effective. Recombinant vitronectin was the only defined functional alternative to Matrigel, supporting sustained self-renewal and pluripotency in three independent hESC lines.
Sigma-Aldrich
Vitronectin human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture