- Characterization of a novel CC chemokine, HCC-4, whose expression is increased by interleukin-10.
Characterization of a novel CC chemokine, HCC-4, whose expression is increased by interleukin-10.
We have identified and characterized a human beta (CC) chemokine, designated HCC-4, that is most closely related to HCC-1 and which demonstrates chemotactic activity for monocytes. Northern analysis of multiple tissue blots and of activated monocytes mRNA shows expression of a 500-bp mRNA. A 1,500-bp mRNA was highly expressed in monocytes activated 12 hours in the presence of interleukin-10 (IL-10) but was absent in monocytes activated for only 1 hour regardless of the presence or absence of IL-10. The upregulation of expression in the presence of IL-10 is in contrast to the downregulatory effects of IL-10 on expression of most other chemokines. Recombinant HCC-4 demonstrated chemotactic activity for human monocytes and THP-1 monocyte cells but not for resting lymphocytes or neutrophils. HCC-4 also induced a Ca2+ flux in THP-1 cells that was desensitized by prior exposure to RANTES. Taken together, these data indicate that HCC-4 is a novel chemokine whose expression is uniquely upregulated by IL-10.