Merck
  • Fiber type composition of epaxial muscles is geared toward facilitating rapid spinal extension in the leaper Galago senegalensis.
Fiber type composition of epaxial muscles is geared toward facilitating rapid spinal extension in the leaper Galago senegalensis.
American journal of physical anthropology (2018-01-11)
Emranul Huq, Andrea B Taylor, Zuowei Su, Christine E Wall
ABSTRACT
We hypothesized that the vertical leaper Galago senegalensis will have epaxial extensor muscles with a fast fiber phenotype to facilitate rapid spinal extension during leaping in comparison to the slow-moving quadruped Nycticebus coucang. To test this, we determined the percentage of fiber cross-sectional area (%CSA) devoted to Type 2 fibers in epaxial muscles of G. senegalensis compared to those of N. coucang. Immunohistochemistry was used to identify Type 1, Type 2, and hybrid fibers in iliocostalis, longissimus, and multifidus muscles of G. senegalensis (n = 3) and N. coucang (n = 3). Serial muscle sections were used to estimate and compare proportions, cross-sectional areas (CSAs), and %CSAs of Type 1, Type 2, and hybrid fibers between species. Epaxial muscles of G. senegalensis were comprised predominantly of Type 2 fibers with large CSAs (%CSA range ≈ 83-94%; range of mean CSA = 1,218-1,586 μm2 ). N. coucang epaxial muscles were comprised predominantly Type 1 fibers with large CSAs (%CSA range ≈ 69-77%; range of mean CSA = 983-1,220 μm2 ). The predominance of Type 2 fibers in G. senegalensis epaxial muscles facilitates rapid muscle excursion and spinal extension during leaping, and is consistent with their relatively long muscle fibers. The predominance of Type 1 fibers in N. coucang epaxial muscles may aid in maintaining stable postures during bridging and cantilevering behaviors characteristic of slow-climbing. These histochemical characteristics highlight the major divergent locomotor repertoires of G. senegalensis and N. coucang.
Sigma-Aldrich
Anti-Myosin (Skeletal, Fast) antibody, Mouse monoclonal, clone MY-32, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Slow) antibody produced in mouse, clone NOQ7.5.4D, ascites fluid