Merck
  • In vitro propagation of Dendrobium macrostachyum Lindl.--a threatened orchid.
In vitro propagation of Dendrobium macrostachyum Lindl.--a threatened orchid.
Indian journal of experimental biology (2003-03-08)
A N Pyati, H N Murthy, E J Hahn, K Y Paek
ABSTRACT
In vitro propagation of Dendrobium macrostachyum, a threatened and endemic species was achieved through nodal explants. The nodal explants were cultured on Murashige and Skoog (MS) basal medium and MS medium supplemented with N6-benzyladenine (BA-2.22, 4.44 and 8.88 microM), Kinetin (KN-2.32, 4.65, and 9.29 microM) and Coconut water (CW, 5, 10 and 15%) individually or in combination with 2.69 microM alpha-naphthalene acetic acid (NAA). Axillary shoots were induced directly from nodal explants in medium containing BA, KN or CW. Optimal shoot induction (6 shoots/explant) was attained from nodal explants cultured on medium supplemented with 15% CW. Well developed shoots rooted at an average 5 roots per shoot in half strength MS medium devoid of any growth regulators.
Sigma-Aldrich
Phytamax Orchid Medium with Charcoal and Banana Powder, powder, suitable for plant cell culture
Sigma-Aldrich
Phytamax Orchid Maintenance Medium, powder, suitable for plant cell culture
Sigma-Aldrich
Phytamax Orchid Maintenance Medium without Charcoal, powder, suitable for plant cell culture
Sigma-Aldrich
Phytamax Orchid Multiplication Medium, powder, suitable for plant cell culture