Merck
  • Occurrence and risk assessment of mycotoxins, acrylamide, and furan in Latvian beer.
Occurrence and risk assessment of mycotoxins, acrylamide, and furan in Latvian beer.
Food additives & contaminants. Part B, Surveillance (2018-03-17)
Estefanija Bogdanova, Irina Rozentale, Iveta Pugajeva, Emeka E Emecheta, Vadims Bartkevics
ABSTRACT
This work reports data on the occurrence of nine mycotoxins and two food processing contaminants - acrylamide and furan - in a total of 100 beers produced in Latvia. Mycotoxins were detected by high-performance liquid chromatography (HPLC) coupled with time-of-flight mass spectrometry, acrylamide by HPLC coupled with quadrupole-Orbitrap mass spectrometry, and furan by headspace gas chromatography-mass spectrometry. The most frequently occurring mycotoxins were HT-2 and deoxynivalenol (DON), which were detected in 52% and 51% of the analysed samples. The highest content was observed for DON, reaching the maximum of 248 µg kg-1. Furan was ubiquitous, and 74% of the samples contained acrylamide. In terms of the estimated exposure, the biggest potential risk was identified for HT-2 representing more than 11% of tolerable weekly intake. The margin of exposure approach indicated the exposure to furan through beer as significant, this parameter being close to the critical limit.
Sigma-Aldrich
Furan-d4, ≥98 atom % D, ≥99% (CP), contains 0.025 wt. % BHT as stabilizer