Merck
  • Pseudouridylation of tRNA-Derived Fragments Steers Translational Control in Stem Cells.
Pseudouridylation of tRNA-Derived Fragments Steers Translational Control in Stem Cells.
Cell (2018-04-10)
Nicola Guzzi, Maciej Cieśla, Phuong Cao Thi Ngoc, Stefan Lang, Sonali Arora, Marios Dimitriou, Kristyna Pimková, Mikael N E Sommarin, Roberto Munita, Michal Lubas, Yiting Lim, Kazuki Okuyama, Shamit Soneji, Göran Karlsson, Jenny Hansson, Göran Jönsson, Anders H Lund, Mikael Sigvardsson, Eva Hellström-Lindberg, Andrew C Hsieh, Cristian Bellodi
ABSTRACT
Pseudouridylation (Ψ) is the most abundant and widespread type of RNA epigenetic modification in living organisms; however, the biological role of Ψ remains poorly understood. Here, we show that a Ψ-driven posttranscriptional program steers translation control to impact stem cell commitment during early embryogenesis. Mechanistically, the Ψ "writer" PUS7 modifies and activates a novel network of tRNA-derived small fragments (tRFs) targeting the translation initiation complex. PUS7 inactivation in embryonic stem cells impairs tRF-mediated translation regulation, leading to increased protein biosynthesis and defective germ layer specification. Remarkably, dysregulation of this posttranscriptional regulatory circuitry impairs hematopoietic stem cell commitment and is common to aggressive subtypes of human myelodysplastic syndromes. Our findings unveil a critical function of Ψ in directing translation control in stem cells with important implications for development and disease.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Nuclease P1 from Penicillium citrinum, lyophilized powder, ≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase)
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Sodium cyanoborodeuteride, 97 atom % D, ≥96% (CP)
Sigma-Aldrich
Giemsa Stain, Modified Solution, according to Giemsa
Sigma-Aldrich
MISSION® esiRNA, targeting human PABPC1
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Formaldehyde-d2 solution, ~20 wt. % in D2O, 98 atom % D
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse