Merck
Dynarrestin, a Novel Inhibitor of Cytoplasmic Dynein.
Cell chemical biology (2018-02-06)
Susanne Höing, Ting-Yu Yeh, Matthias Baumann, Nancy E Martinez, Peter Habenberger, Lea Kremer, Hannes C A Drexler, Philipp Küchler, Peter Reinhardt, Axel Choidas, Mia-Lisa Zischinsky, Gunther Zischinsky, Swaran Nandini, Aaron P Ledray, Stephanie A Ketcham, Lydia Reinhardt, Masin Abo-Rady, Michael Glatza, Stephen J King, Peter Nussbaumer, Slava Ziegler, Bert Klebl, Trina A Schroer, Hans R Schöler, Herbert Waldmann, Jared Sterneckert
ABSTRACT
Aberrant hedgehog (Hh) signaling contributes to the pathogenesis of multiple cancers. Available inhibitors target Smoothened (Smo), which can acquire mutations causing drug resistance. Thus, compounds that inhibit Hh signaling downstream of Smo are urgently needed. We identified dynarrestin, a novel inhibitor of cytoplasmic dyneins 1 and 2. Dynarrestin acts reversibly to inhibit cytoplasmic dynein 1-dependent microtubule binding and motility in vitro without affecting ATP hydrolysis. It rapidly and reversibly inhibits endosome movement in living cells and perturbs mitosis by inducing spindle misorientation and pseudoprometaphase delay. Dynarrestin reversibly inhibits cytoplasmic dynein 2-dependent intraflagellar transport (IFT) of the cargo IFT88 and flux of Smo within cilia without interfering with ciliogenesis and suppresses Hh-dependent proliferation of neuronal precursors and tumor cells. As such, dynarrestin is a valuable tool for probing cytoplasmic dynein-dependent cellular processes and a promising compound for medicinal chemistry programs aimed at development of anti-cancer drugs.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)
Sigma-Aldrich
Dynarrestin, ≥98% (HPLC)
Sigma-Aldrich
Cytoplasmic Dynein Inhibitor, Ciliobrevin D, Ciliobrevin D is a cell-permeable, reversible, and specific blocker of AAA+ ATPase motor cytoplasmic dynein. Disrupts spindle pole focusing and kinetochore-microtubule attachment (~10 to 40 µM).