Merck
  • The immune checkpoint molecule V-set Ig domain-containing 4 is an independent prognostic factor for multiple myeloma.
The immune checkpoint molecule V-set Ig domain-containing 4 is an independent prognostic factor for multiple myeloma.
Oncotarget (2017-09-25)
Jin Roh, Youkyoung Jeon, A-Neum Lee, Sang Min Lee, YeonMee Kim, Chang Ohk Sung, Chan-Jeoung Park, Jung Yong Hong, Dok Hyun Yoon, Cheolwon Suh, Jooryung Huh, Inhak Choi, Chan-Sik Park
ABSTRACT
Multiple myeloma (MM) remains as an incurable disease, despite recent substantial improvements in treatment. Therefore, development of novel biomarkers for risk stratification and new therapeutic targets are imperative. One of the emerging treatments for MM is the immune checkpoint blockades. V-set Ig domain-containing 4 (VSIG4) is a lately studied B7-related immune checkpoint modulator. We assessed the VSIG4 expression in patients with MM and its prognostic impact. We analyzed 81 bone marrow and 66 extramedullary biopsy samples of MM patients using immunohistochemistry. VSIG4 mRNA expression data from the Multiple Myeloma Genomics Portal (MMGP) were analyzed to validate our results. The overall survival (OS) of the high VSIG4 expression group was significantly poorer than that of the low VSIG4 expression group (p = 0.046). VSIG4 expression was remained statistically significant after adjustment for revised international staging system (rISS) and Mayo stratification algorithm (mSMART) risk classification, respectively (p = 0.019 and 0.017). Corroborating results were also observed on analyses of VSIG4 expression in patients with extramedullary MM and external data from the MMGP. Our results suggest that VSIG4 expression in MM is an independent indicator of poor prognosis, implying a possible therapeutic target for immunotherapy for MM.
Sigma-Aldrich
Anti-VSIG4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.