Merck
  • Targeting prohibitins induces apoptosis in acute myeloid leukemia cells.
Targeting prohibitins induces apoptosis in acute myeloid leukemia cells.
Oncotarget (2016-08-20)
Helena Pomares, Claudia M Palmeri, Daniel Iglesias-Serret, Cristina Moncunill-Massaguer, José Saura-Esteller, Sonia Núñez-Vázquez, Enric Gamundi, Montserrat Arnan, Sara Preciado, Fernando Albericio, Rodolfo Lavilla, Gabriel Pons, Eva M González-Barca, Ana M Cosialls, Joan Gil
ABSTRACT
Fluorizoline is a new synthetic molecule that induces apoptosis by selectively targeting prohibitins (PHBs). In this study, the pro-apoptotic effect of fluorizoline was assessed in two cell lines and 21 primary samples from patients with debut of acute myeloid leukemia (AML). Fluorizoline induced apoptosis in AML cells at concentrations in the low micromolar range. All primary samples were sensitive to fluorizoline irrespectively of patients' clinical or genetic features. In addition, fluorizoline inhibited the clonogenic capacity and induced differentiation of AML cells. Fluorizoline increased the mRNA and protein levels of the pro-apoptotic BCL-2 family member NOXA both in cell lines and primary samples analyzed. These results suggest that targeting PHBs could be a new therapeutic strategy for AML.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-MAP Kinase 2/Erk2 Antibody, clone 1B3B9, clone 1B3B9, Upstate®, from mouse
Sigma-Aldrich
Fluorizoline, ≥98% (HPLC)
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®
Sigma-Aldrich
Anti-REA Antibody, Upstate®, from rabbit