Merck
  • Simple paired heavy- and light-chain antibody repertoire sequencing using endoplasmic reticulum microsomes.
Simple paired heavy- and light-chain antibody repertoire sequencing using endoplasmic reticulum microsomes.
Genome medicine (2018-04-29)
Praneeth Reddy Devulapally, Jörg Bürger, Thorsten Mielke, Zoltán Konthur, Hans Lehrach, Marie-Laure Yaspo, Jörn Glökler, Hans-Jörg Warnatz
ABSTRACT
Existing methods for paired antibody heavy- and light-chain repertoire sequencing rely on specialized equipment and are limited by their commercial availability and high costs. Here, we report a novel simple and cost-effective emulsion-based single-cell paired antibody repertoire sequencing method that employs only basic laboratory equipment. We performed a proof-of-concept using mixed mouse hybridoma cells and we also showed that our method can be used for discovery of novel antigen-specific monoclonal antibodies by sequencing human CD19+ B cell IgM and IgG repertoires isolated from peripheral whole blood before and seven days after Td (Tetanus toxoid/Diphtheria toxoid) booster immunization. We anticipate broad applicability of our method for providing insights into adaptive immune responses associated with various diseases, vaccinations, and cancer immunotherapies.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Magnesium acetate solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Span® 80, nonionic surfactant
Sigma-Aldrich
Mineral oil, BioReagent, Molecular Biology, light oil
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
TWEEN® 80, BioXtra, viscous liquid