Merck
  • Simultaneous determination of levofloxacin, gatifloxacin and moxifloxacin in serum by liquid chromatography with column switching.
Simultaneous determination of levofloxacin, gatifloxacin and moxifloxacin in serum by liquid chromatography with column switching.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2004-09-11)
Hoang Anh Nguyen, Jean Grellet, Boubakar B Ba, Claudine Quentin, Marie-Claude Saux
ABSTRACT
Liquid chromatography with a column-switching technique was developed for simultaneous direct quantification of levofloxacin, gatifloxacin and moxifloxacin in human serum. Serum samples were injected on a LiChroCART 4-4 pre-column (PC) filled with a LiChrospher 100 RP-18, 5 microm where fluoroquinolones (FQs) were purified and concentrated. The FQs were back-flushed from the PC and then separated on a Supelcosil ABZ+ Plus (150 mm x 4.6 mm i.d.) analytical column with a mobile phase containing 10 mM phosphate buffer (pH 2.5), acetonitrile (88:12, v/v) and 2mM tetrabutyl ammonium bromide. The effects of ion-pair reagents, buffer type, pH and acetonitrile concentrations in the mobile phase on the separation of the three FQs were investigated. Fluorescence detection provided sufficient sensitivity to achieve a quantification limit of 125 ng/ml for levofloxacin and moxifloxacin; 162.5 ng/ml for gatifloxacin with a 5 microl sample size. The on-line process of extraction avoids time-consuming treatment of the samples before injection and run time is shortened. The recovery, selectivity, linearity, precision and accuracy of the method are convenient for pharmacokinetic studies or routine assays.
Product Description
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL ABZ+Plus Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 3 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
SUPELCOSIL ABZ+Plus Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL ABZ+Plus Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 3 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm