Skip to Content
Merck
ZSTK474 ≥98% (HPLC)

ZSTK474

ZSTK474 ≥98% (HPLC)
Synonym(s):
2-(2-Difluoromethylbenzoimidazol-1-yl)-4, 6-dimorpholino-1, 3, 5-triazine
Empirical Formula (Hill Notation):
C19H21F2N7O2
CAS Number:
Molecular Weight:
417.41

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing