![Zolmitriptan Related Compound H United States Pharmacopeia (USP) Reference Standard, pharmaceutical primary standard](/deepweb/assets/sigmaaldrich/product/structures/107/279/ad608903-72bb-4b5a-a710-ec607eb10505/640/ad608903-72bb-4b5a-a710-ec607eb10505.png)
Zolmitriptan Related Compound H
![Zolmitriptan Related Compound H United States Pharmacopeia (USP) Reference Standard, pharmaceutical primary standard](/deepweb/assets/sigmaaldrich/product/structures/107/279/ad608903-72bb-4b5a-a710-ec607eb10505/640/ad608903-72bb-4b5a-a710-ec607eb10505.png)
Synonym(s):
4,4-Diethoxy-N,N-dimethyl-1-butanamine, 1-(N,N-Dimethylamino)-4,4-diethoxybutane, 4,4-Diethoxy-N,N-dimethylbutanamine, 4,4-Diethoxy-N,N-dimethylbutylamine, 4-Dimethylaminobutanal diethyl acetal, N,N-Dimethyl-4-aminobutanal diethyl acetal
Empirical Formula (Hill Notation):
C10H23NO2
CAS Number:
Molecular Weight:
189.30
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing