![Zolmitriptan Related Compound G United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/335/192/2066b49d-e1df-4844-a6b5-69dcdf04e2a9/640/2066b49d-e1df-4844-a6b5-69dcdf04e2a9.png)
Zolmitriptan Related Compound G
![Zolmitriptan Related Compound G United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/335/192/2066b49d-e1df-4844-a6b5-69dcdf04e2a9/640/2066b49d-e1df-4844-a6b5-69dcdf04e2a9.png)
Synonym(s):
(S)-4-(4-Aminobenzyl)-2(1H)-oxazolidinone, (4S)-4-[(4-Aminophenyl)methyl]-2-oxazolidinone
Empirical Formula (Hill Notation):
C10H12N2O2
CAS Number:
Molecular Weight:
192.21
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing