![Zirconium(IV) oxide-yttria stabilized nanopowder](/deepweb/assets/sigmaaldrich/product/structures/862/190/5b8199f1-e7a8-4881-afec-1957b843e9ed/640/5b8199f1-e7a8-4881-afec-1957b843e9ed.png)
Zirconium(IV) oxide-yttria stabilized
![Zirconium(IV) oxide-yttria stabilized nanopowder](/deepweb/assets/sigmaaldrich/product/structures/862/190/5b8199f1-e7a8-4881-afec-1957b843e9ed/640/5b8199f1-e7a8-4881-afec-1957b843e9ed.png)
Synonym(s):
YSZ, Yttria stabilized zirconia, Yttrium stabilized zirconium oxide
Linear Formula:
ZrO2 · Y2O3
CAS Number:
Molecular Weight:
349.03
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-2 of 2