![Ziprasidone Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/251/438/17986f3e-5440-4614-aacc-2d5045c839df/640/17986f3e-5440-4614-aacc-2d5045c839df.jpg)
Ziprasidone Related Compound A
![Ziprasidone Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/251/438/17986f3e-5440-4614-aacc-2d5045c839df/640/17986f3e-5440-4614-aacc-2d5045c839df.jpg)
Synonym(s):
3-(Piperazin-1-yl)benzo[d]isothiazole monohydrochloride
Empirical Formula (Hill Notation):
C11H13N3S · HCl
CAS Number:
Molecular Weight:
255.77
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing