Vitexin-2"-O-rhamnoside
Synonym(s):
Vitexin 2-O-rhamnoside, 2-O-Rhamnosylvitexin
Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
Products
Formula Weight
Manufacturer
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing