![Vincamine for system suitability CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/811/198/1f01ac47-3d9d-44d7-8887-e468eaceef04/640/1f01ac47-3d9d-44d7-8887-e468eaceef04.png)
Vincamine for system suitability
![Vincamine for system suitability CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/811/198/1f01ac47-3d9d-44d7-8887-e468eaceef04/640/1f01ac47-3d9d-44d7-8887-e468eaceef04.png)
Synonym(s):
Vincamine
Empirical Formula (Hill Notation):
C21H26N2O3
CAS Number:
Molecular Weight:
354.44
Products
Formula Weight
Manufacturer
Melting Point (°C)
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing