Skip to Content
Merck
Undecanoic acid analytical standard

Undecanoic acid

Undecanoic acid analytical standard
Synonym(s):
Hendecanoic acid
Linear Formula:
CH3(CH2)9COOH
CAS Number:
Molecular Weight:
186.29
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing