
Ubidecarenone impurity D

Synonym(s):
Coenzyme Q9, Q-9, Ubiquinone-45, Ubiquinone-9
Empirical Formula (Hill Notation):
C54H82O4
CAS Number:
Molecular Weight:
795.23
Products
Product Category
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing