Skip to Content
Merck
Tungsten(VI) chloride powder, ≥99.99% trace metals basis (purity excludes molybdenum)

Tungsten(VI) chloride

Tungsten(VI) chloride powder, ≥99.99% trace metals basis (purity excludes molybdenum)
Synonym(s):
Tungsten hexachloride
Linear Formula:
WCl6
CAS Number:
Molecular Weight:
396.56
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core