Skip to Content
Merck
TSKgel® Phenyl-5PW (30) Bulk Media phase phenyl, bottle of 25 mL, 30 μm particle size

TSKgel® Phenyl-5PW (30) Bulk Media

TSKgel® Phenyl-5PW (30) Bulk Media phase phenyl, bottle of 25 mL, 30 μm particle size
Synonym(s):
TSKgel® Phenyl-5PW HIC Bulk Media

Products

Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Technique