![TSKgel® G6000HXL HPLC Column phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 9 μm particle size](/deepweb/assets/sigmaaldrich/product/images/427/034/89541ec9-8bb8-417b-a64d-8bccc3bee58b/640/89541ec9-8bb8-417b-a64d-8bccc3bee58b.jpg)
TSKgel® G6000HXL (9 µm) HPLC Columns
![TSKgel® G6000HXL HPLC Column phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 9 μm particle size](/deepweb/assets/sigmaaldrich/product/images/427/034/89541ec9-8bb8-417b-a64d-8bccc3bee58b/640/89541ec9-8bb8-417b-a64d-8bccc3bee58b.jpg)
Synonym(s):
TSKgel® G1000HXL HPLC Column for Light Scattering Applications
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing