Tropicamide
Synonym(s):
N-Ethyl-2-phenyl-N-(4-pyridylmethyl)hydracrylamide, Ro 1-7683, Tropicamide
Empirical Formula (Hill Notation):
C17H20N2O2
CAS Number:
Molecular Weight:
284.35
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing