Skip to Content
Merck
Tri(propylene glycol) methyl ether, mixture of isomers 96%

Tri(propylene glycol) methyl ether, mixture of isomers

Tri(propylene glycol) methyl ether, mixture of isomers 96%
Linear Formula:
CH3(OC3H6)3OH
CAS Number:
Molecular Weight:
206.28
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Polymer Composition
Polymer Type
Showing 1-2 of 2