![Tripropionin ≥97.1%, FCC, FG](/deepweb/assets/sigmaaldrich/product/structures/213/784/5924cd38-7732-426f-b0ef-d5709b1835db/640/5924cd38-7732-426f-b0ef-d5709b1835db.png)
Tripropionin
![Tripropionin ≥97.1%, FCC, FG](/deepweb/assets/sigmaaldrich/product/structures/213/784/5924cd38-7732-426f-b0ef-d5709b1835db/640/5924cd38-7732-426f-b0ef-d5709b1835db.png)
Synonym(s):
Glyceryl tripropionate
Linear Formula:
(C2H5CO2CH2)2CH(O2CC2H5)
CAS Number:
Molecular Weight:
260.28
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing